Learn More
Biomatik Corporation Mouse Thrombospondin 1 (THBS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07641
Encompass_Preferred
Mouse Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.266ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35441. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3916