Learn More
Biomatik Corporation Rat Thrombospondin 1 (THBS1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07642
Encompass_Preferred
Rat Thrombospondin 1 (THBS1) ELISA Kit for detecting THBS1 in plasma. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q71SA3. Gene ID: 445442. Target Synonyms: TSP1; Thrombospondin-1p180. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3917