Learn More
Biomatik Corporation Mouse Thrombospondin 2 (THBS2) ELISA Kit, 96 tests
Catalog No. 501503918
Encompass_Preferred
Mouse Thrombospondin 2 (THBS2) ELISA Kit for detecting THBS2 in plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03350. Gene ID: 21826. Target Synonyms: TSP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3918 Supplier Biomatik Corporation Supplier No. EKU07644
Mouse Thrombospondin 2 (THBS2) ELISA Kit for detecting THBS2 in plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.135ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q03350. Gene ID: 21826. Target Synonyms: TSP2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.