Learn More
Biomatik Corporation Mouse Thrombospondin 4 (THBS4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07648
Encompass_Preferred
Mouse Thrombospondin 4 (THBS4) ELISA Kit for detecting THBS4 in plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z1T2. Gene ID: 21828. Target Synonyms: TSP4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3920