Learn More
Biomatik Corporation Human Thy1 Cell Surface Antigen (Thy1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07652
Encompass_Preferred
Human Cluster of Differentiation 90 (CD90) ELISA Kit for detecting CD90 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.50-800pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04216. Gene ID: 7070. Target Synonyms: Thy1; CDw90; THY mocyte differentiation antigen 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3922