Learn More
Biomatik Corporation Mouse Thyroglobulin (TG) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07673
Encompass_Preferred
Mouse Thyroglobulin (TG) ELISA Kit for detecting TG in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O08710. Gene ID: 21819. Target Synonyms: AITD3; TGN. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3931