Learn More
Biomatik Corporation Dog Thyroid Stimulating Hormone (TSH) ELISA Kit, 96 tests
Catalog No. 501503932
Encompass_Preferred
The Dog Thyroid Stimulating Hormone (TSH) ELISA Kit allows for quantitative determination of Thyroid Stimulating Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 185.2-15,000 pg/mL. Sensitivity: 72.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Thyroid Stimulating Hormone and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3932 Supplier Biomatik Corporation Supplier No. EKU07682
The Dog Thyroid Stimulating Hormone (TSH) ELISA Kit allows for quantitative determination of Thyroid Stimulating Hormone in Serum, Plasma and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 185.2-15,000 pg/mL. Sensitivity: 72.4 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Thyroid Stimulating Hormone and analogues was observed. Restriction: For Research Use Only.