Learn More
Biomatik Corporation Mouse Thyroid Stimulating Hormone (TSH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07683
Encompass_Preferred
Mouse Thyroid Stimulating Hormone (TSH) ELISA Kit for detecting TSH in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 6.4-4,000pg/ml. Sensitivity: 2.5pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01216; P12656. Gene ID: 12640. Target Synonyms: Thyrotropin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3933