Learn More
Biomatik Corporation Human Thyrotropin Releasing Hormone (TRH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07687
Encompass_Preferred
Human Thyrotropin Releasing Hormone (TRH) ELISA Kit for detecting TRH in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.17-500pg/ml. Sensitivity: 2.94pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P20396. Gene ID: 7200. Target Synonyms: TRF; Thyrotropin-Releasing Factor; Thyroliberin; Protirelin; Pro-tirelin; Prothyroliberin; TSH-releasing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3935