Learn More
Biomatik Corporation Mouse Tissue Factor (TF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07697
Encompass_Preferred
Mouse Tissue Factor (TF) ELISA Kit for detecting TF in plasma. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.6pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20352. Gene ID: 14066. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3942