Learn More
Biomatik Corporation Actin-α-1 Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50015
Encompass_Preferred
Actin-α-1 Colorimetric Cell-Based ELISA Kit for detecting Actin-α-1 in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P68133. Gene ID: 58. Target Synonyms: ACTA; ACTA1; ACTS; Actin alpha skeletal muscle; Alpha-actin 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8893