Learn More
Biomatik Corporation Rat Tissue Factor (TF) ELISA Kit, 96 tests
Catalog No. 501503943
Encompass_Preferred
Rat Tissue Factor (TF) ELISA Kit for detecting TF in plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42533. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3943 Supplier Biomatik Corporation Supplier No. EKU07699
Rat Tissue Factor (TF) ELISA Kit for detecting TF in plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.4pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42533. Target Synonyms: CD142; FIII; F3; TFA; Thromboplastin; Coagulation Factor III. Restrictions: For Research Use Only. Not for use in diagnostic procedures.