Learn More
Biomatik Corporation Rabbit Tissue Factor Pathway Inhibitor (TFPI) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07704
Encompass_Preferred
Rabbit Tissue Factor Pathway Inhibitor (TFPI) ELISA Kit for detecting TFPI in plasma. Species Reactivity: Rabbit (Oryctolagus cuniculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19761. Gene ID: 100009401. Target Synonyms: TFI; EPI; LACI; TFPI1; Lipoprotein-Associated Coagulation Inhibitor; Extrinsic pathway inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3947