Learn More
Biomatik Corporation Horse Tissue Inhibitors of Metalloproteinase 1 (TIMP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07708
Encompass_Preferred
The Horse Tissue Inhibitors of Metalloproteinase 1 (TIMP1) ELISA Kit allows for quantitative determination of Tissue Inhibitors of Metalloproteinase 1 in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Equus caballus; Equine (Horse). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.241 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O02722. Cross Activity: No significant cross-reactivity or interference between Tissue Inhibitors of Metalloproteinase 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3949