Learn More
Biomatik Corporation Estrogen Receptor-β Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50212
Encompass_Preferred
Estrogen Receptor-β Colorimetric Cell-Based ELISA Kit for detecting Estrogen Receptor-β in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q92731. Gene ID: 2100. Target Synonyms: ESR2; ESTRB; Estrogen receptor beta; NR3A2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8924