Learn More
Biomatik Corporation Rat Tissue Inhibitors of Metalloproteinase 1 (TIMP1) ELISA Kit, 96 tests
Catalog No. 501503952
Encompass_Preferred
Rat Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit for detecting TIMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30120. Gene ID: 116510. Target Synonyms: EPO; CLGI; EPA; HCI; Erythroid Potentiating Activity; Collagenase Inhibitor; Metalloproteinase inhibitor 1; Fibroblast collagenase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3952 Supplier Biomatik Corporation Supplier No. EKU07712
Rat Tissue Inhibitors Of Metalloproteinase 1 (TIMP1) ELISA Kit for detecting TIMP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.067ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30120. Gene ID: 116510. Target Synonyms: EPO; CLGI; EPA; HCI; Erythroid Potentiating Activity; Collagenase Inhibitor; Metalloproteinase inhibitor 1; Fibroblast collagenase inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.