Learn More
Biomatik Corporation Human Tissue Inhibitors of Metalloproteinase 2 (TIMP2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07716
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.94-60ng/ml. Sensitivity: 0.41ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16035. Gene ID: 7077. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3955