Learn More
Biomatik Corporation Mouse Tissue Inhibitors of Metalloproteinase 2 (TIMP2) ELISA Kit, 96 tests
Catalog No. 501503956
Encompass_Preferred
Mouse Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.58ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25785. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3956 Supplier Biomatik Corporation Supplier No. EKU07717
Mouse Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.58ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P25785. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.