Learn More
Biomatik Corporation MITF Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50384
Encompass_Preferred
MITF Colorimetric Cell-Based ELISA Kit for detecting MITF in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O75030. Gene ID: 4286. Target Synonyms: MITF M1; Microphthalmia-associated transcription factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8944