Learn More
Biomatik Corporation Human Tissue Inhibitors of Metalloproteinase 3 (TIMP3) ELISA Kit, 96 tests
Catalog No. 501503958
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35625. Gene ID: 7078. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3958 Supplier Biomatik Corporation Supplier No. EKU07724
Human Tissue Inhibitors Of Metalloproteinase 3 (TIMP3) ELISA Kit for detecting TIMP3 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35625. Gene ID: 7078. Target Synonyms: SFD; TIMP Metallopeptidase Inhibitor 3; Metallopeptidase Inhibitor 3; Sorsby Fundus Dystrophy; Pseudoinflammatory. Restrictions: For Research Use Only. Not for use in diagnostic procedures.