Learn More
Biomatik Corporation Human Toll Like Receptor 10 (TLR10) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07746
Encompass_Preferred
Human Toll Like Receptor 10 (TLR10) ELISA Kit for detecting TLR10 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9BXR5. Gene ID: 81793. Target Synonyms: CD290. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3962