Learn More
Biomatik Corporation Human Toll Like Receptor 2 (TLR2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07747
Encompass_Preferred
Human Toll Like Receptor 2 (TLR2) ELISA Kit for detecting TLR2 in saliva, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.112ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60603. Gene ID: 7097. Target Synonyms: CD282; TIL4; Toll/interleukin-1 receptor-like protein 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3963