Learn More
Biomatik Corporation p16 INK Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50436
Encompass_Preferred
p16 INK Colorimetric Cell-Based ELISA Kit for detecting p16 INK in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P42771. Gene ID: 1029. Target Synonyms: CD2A1; CDK4I; CDKN2; CDKN2A; CDN2; Cyclin-dependent kinase 4 inhibitor A; cyclin-dependent kinase inhibitor 2A; MTS1; Multiple tumor suppressor 1; p14ARF; p16(INK4a) p16-INK4; P16INK4A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8957