Learn More
Biomatik Corporation PEK/PERK Colorimetric Cell-Based ELISA Kit
Supplier:  Biomatik Corporation EKA50471
Encompass_Preferred
PEK/PERK Colorimetric Cell-Based ELISA Kit for detecting PEK/PERK in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q9NZJ5. Gene ID: 9451. Target Synonyms: E2AK3; EIF2AK3; Eukaryotic translation initiation factor 2-alpha kinase 3 precursor; HsPEK; PEK; PERK; PRKR-like endoplasmic reticulum kinase; Pancreatic eIF2-alpha kinase; kinase PERK. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8959