Learn More
Biomatik Corporation Cattle Toll Like Receptor 4 (TLR4) ELISA Kit, 96 tests
Catalog No. 501503965
Encompass_Preferred
The Cattle Toll Like Receptor 4 (TLR4) ELISA Kit allows for quantitative determination of Toll Like Receptor 4 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.236 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9GL65. Cross Activity: No significant cross-reactivity or interference between Toll Like Receptor 4 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3965 Supplier Biomatik Corporation Supplier No. EKU07752
The Cattle Toll Like Receptor 4 (TLR4) ELISA Kit allows for quantitative determination of Toll Like Receptor 4 in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.236 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q9GL65. Cross Activity: No significant cross-reactivity or interference between Toll Like Receptor 4 and analogues was observed. Restriction: For Research Use Only.