Learn More
Biomatik Corporation Mouse Toll Like Receptor 4 (TLR4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07754
Encompass_Preferred
Mouse Toll Like Receptor 4 (TLR4) ELISA Kit for detecting TLR4 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9QUK6. Gene ID: 21898. Target Synonyms: CD284; TOLL; HToll. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3967