Learn More
Biomatik Corporation Human Toll Like Receptor 5 (TLR5) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07756
Encompass_Preferred
Human Toll Like Receptor 5 (TLR5) ELISA Kit for detecting TLR5 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.30ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O60602. Gene ID: 7100. Target Synonyms: CD285; SLEB1; TIL3; Toll/Interleukin-1 Receptor-Like Protein 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3969