Learn More
Biomatik Corporation Human Toll Like Receptor 6 (TLR6) ELISA Kit, 96 tests
Catalog No. 501503970
Encompass_Preferred
Human Toll Like Receptor 6 (TLR6) ELISA Kit for detecting TLR6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2C9. Gene ID: 10333. Target Synonyms: CD286. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3970 Supplier Biomatik Corporation Supplier No. EKU07757
Human Toll Like Receptor 6 (TLR6) ELISA Kit for detecting TLR6 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.127ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Y2C9. Gene ID: 10333. Target Synonyms: CD286. Restrictions: For Research Use Only. Not for use in diagnostic procedures.