Learn More
Biomatik Corporation BD-2 (Human) Sandwich ELISA Kit
Supplier:  Biomatik Corporation EKA51819
Encompass_Preferred
BD-2 (Human) Sandwich ELISA Kit for detecting BD-2 in plasma, serum, lysate. Species Reactivity: Human (Homo sapiens). Detection Range: 8-1000pg/ml. Sensitivity: . Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: O15263. Gene ID: 100289462; 1673. Target Synonyms: BD-2; hBD-2; Defensin beta 2; Skin-antimicrobial peptide 1; SAP1; DEFB4; DEFB102; DEFB2; DEFB4P; Beta-defensin 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-9103