Learn More
Biomatik Corporation Human Topoisomerase II Beta (TOP2b) ELISA Kit, 96 tests
Catalog No. 501503978
Encompass_Preferred
Human Topoisomerase II Beta (TOP2b) ELISA Kit for detecting TOP2b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02880. Gene ID: 7155. Target Synonyms: TOPIIB; Top2beta; DNA topoisomerase II, beta isozyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3978 Supplier Biomatik Corporation Supplier No. EKU07771
Human Topoisomerase II Beta (TOP2b) ELISA Kit for detecting TOP2b in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q02880. Gene ID: 7155. Target Synonyms: TOPIIB; Top2beta; DNA topoisomerase II, beta isozyme. Restrictions: For Research Use Only. Not for use in diagnostic procedures.