Learn More
Biomatik Corporation Human Transcobalamin I (TCN1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07776
Encompass_Preferred
Human Transcobalamin I (TCN1) ELISA Kit for detecting TCN1 in serum, plasma, saliva, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.0pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20061. Gene ID: 6947. Target Synonyms: TCI; TC1; Haptocorin; Haptocorrin; Vitamin B12 Binding Protein,R Binder Family. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3979