Learn More
Biomatik Corporation Pig Transferrin (TRF) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07785
Encompass_Preferred
Pig Transferrin (TF) ELISA Kit for detecting TF in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.147ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P09571. Target Synonyms: TRF; Siderophilin; Serotransferrin; Beta-1 metal-binding globulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3982