Learn More
Biomatik Corporation Mouse Transferrin Receptor (TFR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07789
Encompass_Preferred
Mouse Transferrin Receptor (TFR) ELISA Kit for detecting TFR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q62351. Gene ID: 22042. Target Synonyms: CD71; TFR1; P90; TFRC; T9; Trfr; sTfR; Transferrin receptor protein 1, serum form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3984