Learn More
Biomatik Corporation Rat Transferrin Receptor (TFR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07790
Encompass_Preferred
Rat Transferrin Receptor (TFR) ELISA Kit for detecting TFR in serum, plasma, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.239ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99376. Gene ID: 64678. Target Synonyms: CD71; TFR1; P90; TFRC; T9; Trfr; sTfR; Transferrin receptor protein 1, serum form. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3985