Learn More
Biomatik Corporation Human Transforming Growth Factor Alpha (TGFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07794
Encompass_Preferred
Human Transforming Growth Factor Alpha (TGFa) ELISA Kit for detecting TGFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01135. Gene ID: 7039. Target Synonyms: ETGF; TGF-A; Protransforming Growth Factor alpha; EGF-Like TGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3986