Learn More
Biomatik Corporation Mouse N-Acetyl Beta-D-Glucosaminidase (NAGase) ELISA Kit
Supplier:  Biomatik Corporation EKL59290
Encompass_Preferred
Sample Type: urine.. Species Reactivity: Mouse. Detection Range: 0.781-50ng/mL. Sensitivity: 0.50ng/mL. Format: 96-Well Plate. Assay Type: sandwich. Detection Method: sandwich. Assay Time: 3 hours. UniProt ID: Q9EQQ9. Target Synonyms: MGEA5,MEA5,NCOAT,Hexosaminidase C,N-Acetylhexosaminidase,Hyaluronidase,Meningioma-Expressed Antigen 5,Nuclear Cytoplasmic O-GlcNAcase And Acetyltransferase. Restriction: For Research Use Only.
Catalog No. 50-228-8744