Learn More
Biomatik Corporation Mouse Transforming Growth Factor Alpha (TGFa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07795
Encompass_Preferred
Mouse Transforming Growth Factor Alpha (TGFa) ELISA Kit for detecting TGFa in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 7.8-500pg/ml. Sensitivity: 2.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48030. Gene ID: 21802. Target Synonyms: ETGF; TGF-A; Protransforming Growth Factor alpha; EGF-Like TGF. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3987