Learn More
Biomatik Corporation Dog Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit, 96 tests
Catalog No. 501503990
Encompass_Preferred
The Dog Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit allows for quantitative determination of Transforming Growth Factor Beta 1 in Serum, Platelet-poor Plasma, Tissue homogenates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P54831. Cross Activity: No significant cross-reactivity or interference between Transforming Growth Factor Beta 1 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3990 Supplier Biomatik Corporation Supplier No. EKU07798
The Dog Transforming Growth Factor Beta 1 (TGFb1) ELISA Kit allows for quantitative determination of Transforming Growth Factor Beta 1 in Serum, Platelet-poor Plasma, Tissue homogenates, Cell culture supernates and other biological fluids. Species Reactivity: Canis familiaris; Canine (Dog). Detection Range: 15.6-1000 pg/mL. Sensitivity: 5.7 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P54831. Cross Activity: No significant cross-reactivity or interference between Transforming Growth Factor Beta 1 and analogues was observed. Restriction: For Research Use Only.