Learn More
Biomatik Corporation Cattle Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07808
Encompass_Preferred
The Cattle Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit allows for quantitative determination of Transforming Growth Factor Beta 2 in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Bos taurus; Bovine (Cattle). Detection Range: 15.6-1000 pg/mL. Sensitivity: 6.1 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P21214. Cross Activity: No significant cross-reactivity or interference between Transforming Growth Factor Beta 2 and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-3996