Learn More
Biomatik Corporation Rat Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07812
Encompass_Preferred
Rat Transforming Growth Factor Beta 2 (TGFb2) ELISA Kit for detecting TGFb2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07257. Gene ID: 81809. Target Synonyms: TGF-B2; TGF-Beta2; G-TSF; LAP; Cetermin; Polyergin; Latency-associated peptide; BSC-1 cell growth inhibitor; Glioblastoma-derived T-cell suppressor factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-3997