Learn More
Biomatik Corporation Human Stem Cell Factor (SCF) ELISA Kit
Supplier:  Biomatik Corporation EKL60467
Encompass_Preferred
Human Stem Cell Factor (SCF) ELISA Kit for detecting SCF in serum, plasma, tissue homogenates, cell lysates, cell culture supernatants, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2000pg/ml. Sensitivity: 12.2pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P21583. Gene ID: 4254. Target Synonyms: KITLG; MGF; SF; KL1; Kitl; FPH2; Kit-Ligand; Steel Factor; Mast Cell Growth Factor; Steel factor; Familial Progressive Hyperpigmentation 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8779