Learn More
Biomatik Corporation Mouse Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07816
Encompass_Preferred
Mouse Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P17125. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4000