Learn More
Biomatik Corporation Rat Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit, 96 tests
Catalog No. 501504001
Encompass_Preferred
Rat Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07258. Gene ID: 25717. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4001 Supplier Biomatik Corporation Supplier No. EKU07818
Rat Transforming Growth Factor Beta 3 (TGFb3) ELISA Kit for detecting TGFb3 in serum, plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07258. Gene ID: 25717. Target Synonyms: TGF-B3; LAP; Latency-associated peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.