Learn More
Biomatik Corporation Human Transgelin (TAGLN) ELISA Kit, 96 tests
Catalog No. 501504003
Encompass_Preferred
Human Transgelin (TAGLN) ELISA Kit for detecting TAGLN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01995. Gene ID: 6876. Target Synonyms: SMCC; SM22; TAGLN1; WS3-10; Transgelin Variant 2; 22 kDa actin-binding protein; Protein WS3-10; Smooth muscle protein 22-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4003 Supplier Biomatik Corporation Supplier No. EKU07826
Human Transgelin (TAGLN) ELISA Kit for detecting TAGLN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.060ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q01995. Gene ID: 6876. Target Synonyms: SMCC; SM22; TAGLN1; WS3-10; Transgelin Variant 2; 22 kDa actin-binding protein; Protein WS3-10; Smooth muscle protein 22-alpha. Restrictions: For Research Use Only. Not for use in diagnostic procedures.