Learn More
Biomatik Corporation Human Transglutaminase 1, Keratinocyte (TGM1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07829
Encompass_Preferred
Human Transglutaminase 1 (TGM1) ELISA Kit for detecting TGM1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P22735. Gene ID: 7051. Target Synonyms: TGK; ICR2; KTG; LI; LI1; TGASE; Transglutaminase 1, Keratinocyte; Transglutaminase K; K Polypeptide Epidermal Type I; Protein-Glutamine-Gamma-Glutamyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4005