Learn More
Biomatik Corporation Human Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit
Supplier:  Biomatik Corporation EKL61041
Encompass_Preferred
Human Tissue Inhibitors Of Metalloproteinase 2 (TIMP2) ELISA Kit for detecting TIMP2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 46.8-3000pg/ml. Sensitivity: 14.5pg/ml. Format: 96-Well Plate. Assay Type: Sandwich. Assay Time: 3h. Accession Number: P16035. Gene ID: 7077. Target Synonyms: CSC-21K; TIMP Metallopeptidase Inhibitor 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-8796