Learn More
Biomatik Corporation Rat Transglutaminase 2, Tissue (TGM2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07832
Encompass_Preferred
Rat Transglutaminase 2 (TGM2) ELISA Kit for detecting TGM2 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q6P6R6. Gene ID: 56083. Target Synonyms: tTG; TG2; C Polypeptide,Protein-Glutamine-Gamma-Glutamyltransferase; Transglutaminase 2, Tissue; Transglutaminase C; Transglutaminase H. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4007