Learn More
Biomatik Corporation Human Transglutaminase 3, Epidermal (TGM3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07833
Encompass_Preferred
Human Transglutaminase 3, Epidermal (TGM3) ELISA Kit for detecting TGM3 in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08188. Target Synonyms: TGE; Egt; Protein-Glutamine Gamma-Glutamyltransferase E; Transglutaminase E. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4008