Learn More
Biomatik Corporation Human Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU07844
Encompass_Preferred
Human Transient Receptor Potential Cation Channel Subfamily V, Member 1 (TRPV1) ELISA Kit for detecting TRPV1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.121ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8NER1. Gene ID: 7442. Target Synonyms: VR1; OTRPC1; Vanilloid Receptor Subtype 1; Capsaicin receptor; Osm-9-like TRP channel 1; Vanilloid receptor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4011