Learn More
Biomatik Corporation Human Translocator Protein (TSPO) ELISA Kit, 96 tests
Catalog No. 501504014
Encompass_Preferred
Human Translocator Protein (TSPO) ELISA Kit for detecting TSPO in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30536. Gene ID: 706. Target Synonyms: IBP; PKBS; PBR; pk18; mDRC; MBR; BZRP; DBI; PTBR; Mitochondrial benzodiazepine receptor; Peripheral-Type Benzodiazepine Receptor/Recognition Site. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-4014 Supplier Biomatik Corporation Supplier No. EKU07853
Human Translocator Protein (TSPO) ELISA Kit for detecting TSPO in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.114ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P30536. Gene ID: 706. Target Synonyms: IBP; PKBS; PBR; pk18; mDRC; MBR; BZRP; DBI; PTBR; Mitochondrial benzodiazepine receptor; Peripheral-Type Benzodiazepine Receptor/Recognition Site. Restrictions: For Research Use Only. Not for use in diagnostic procedures.